Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 11-Ketotestosterone Competitive ELISA Kit

Catalog No. EIA11K
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIA11K 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIA11K Supplier Invitrogen™ Supplier No. EIA11K
Add to Cart
Add to Cart

ELISA

The multispecies 11-Ketotestosterone ELISA quantitates 11-Ketotestosterone in extracted serum, plasma, and dried fecal samples, or urine. Principle of the method: The 11-Ketotestosterone Competitive ELISA research-use-only kit is designed to quantitatively measure 11-Ketotestosterone independent of species. A 11-Ketotestosterone standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with goat anti-rabbit IgG antibodies. 11-Ketotestosterone polyclonal rabbit antibodies and 11-Ketotestosterone-peroxidase conjugate is added to the wells. The 11-Ketotestosterone conjugate and any 11-Ketotestosterone in the sample will compete to bind to the 11-Ketotestosterone polyclonal rabbit antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound 11-Ketotestosteron-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of 11-Ketotestosterone present in the sample. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Androgenic hormones, such as testosterone, affect many male organisms' growth, size, and reproduction. In teleost fish, along with testosterone, 11-ketotestosterone plays a significant role by inducing reproductive characteristics in both males and females. In particular male fish, 11-ketotestosterone levels increase during spermatogenesis in the spawning season, while in some female fish, 11-ketotestosterone increases before yolk deposition to regulate ovarian development. The presence and involvement of 11-ketotestosterone in other species, such as humans, have only recently been established. In contrast to fish, primate serum 11-ketotestosterone concentrations were not significantly different in males and females, despite males having significantly higher circulating testosterone. This evidence suggests that 11-ketotestosterone production in these species may not be testis-dependent and primarily originates from adrenal-derived 11-oxyandrogen precursors. Recent studies have discovered more 11-ketotestosterone than its precursors, androstenedione, and testosterone, in prepubertal children and postmenopausal women compared to men. This discovery highlights the potential use of 11-ketotestosterone as a clinical biomarker to screen adrenal androgen excess in disease conditions such as polycystic ovary syndrome (PCOS) and hirsutism in women.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 8.2-2000 pg/mL
Assay Sensitivity 1.85 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Chromone
Sample Type Fecal Extract, Plasma, Serum, Urine
Target Species Chemical
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 11KT; 11-KT11KT; 11-KT;
Interassay CV <8%
Intraassay CV <6%
Kit Contents Pre-coated 96 well plate, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases 17-Hydroxyandrost-4-ene-3,11-dione,11-Ketotestosterone, 11 Oxotestosterone
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Urine 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements 2-8°C
Target 11-Ketotestosterone
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.