Learn More
Arbor Assays 11-Ketotestosterone ELISA Kit, (5 plate)
SDP
Supplier:  Arbor Assays K079H5
Encompass_Preferred
The DetectX 11-Ketotestosterone Immunoassay Kit uses a specifically generated antibody to measure 11-ketotestosterone in urine or in extracted fecal, serum, and plasma samples.
Catalog No. 50-227-8520