Learn More
Medchemexpress LLC 11-Maleimidoundecanoic acid | 57079-01-3 | 99.9% | 25 G

Supplier: Medchemexpress LLC 25GHY130908
11-Maleimidoundecanoic acid is an intermediate utilized for ester and amide linked maleimide monomers. It can also be employed to generate copolymers.
- Appearance: White to off-white solid
- Molecular weight: 281.35
- Molecular formula: C15H23NO4
- Purity: 99.92%
- For research use only
- Storage as powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.