Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 11-Mercapto-1-undecanol

Catalog No. 4475281G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-752-81G 1 g
44-752-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-752-81G Supplier Sigma Aldrich Supplier No. 4475281G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

11-Mercapto-1-undecanol

Specifications

CAS 73768-94-2
Melting Point 33°C to 37°C
Molecular Formula C11H24OS
Linear Formula HS(CH2)11 OH
Quantity 1 g
MDL Number MFCD00239433
Synonym 11-Hydroxy-1-undecanethiol; 11-Hydroxy-1-undecanthiol; 11-Hydroxyundecane-1-thiol; 11-Mercaptoundecanol; MUD
Molecular Weight (g/mol) 204.37
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.