Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 11-Mercaptoundecylphosphoric acid

Catalog No. 67431150MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-431-150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-431-150MG Supplier Sigma Aldrich Supplier No. 67431150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

11-Mercaptoundecylphosphoric acid

Specifications

CAS 188678-49-1
Melting Point 48°C to 52°C
Molecular Formula C11H25O4PS
Linear Formula C11H25O4PS
Quantity 50 mg
MDL Number MFCD09836216
Synonym 11-Mercapto-1-undecyl dihydrogen phosphate; MDPA
Molecular Weight (g/mol) 284.35
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.