Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 11-Methylundecanoatephosphonic acid

Catalog No. 503125967 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5967 250 mg
50-312-5533 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-5967 Supplier Sigma Aldrich Supplier No. SIK750310250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thanks to the carbonyl function, this coupling agent can react with amine or aminooxy function on biomolecules to form imine or oxime ligation.Product of SiKEMIA

Specifications

CAS 83905-96-8
Molecular Formula C12H25O5P
Linear Formula C12H25O5P
Quantity 250 mg
Molecular Weight (g/mol) 280.3
Percent Purity ≥95%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.