Learn More
Perkin Elmer US LLC 11 mm, Hand Crimper Tool
Catalog No. 502104119
Encompass_Preferred
Hand Crimper for manual crimping of 11mm aluminum crimp seals to vials. Crimp capping is still a preferred method by some laboratories to ensure sample integrity, particularly those containing volatile compounds.
Catalog No. 50-210-4119 Supplier Perkin Elmer US LLC Supplier No. 00090699
Hand Crimper for manual crimping of 11mm aluminum crimp seals to vials. Crimp capping is still a preferred method by some laboratories to ensure sample integrity, particularly those containing volatile compounds.