Learn More
Perkin Elmer US LLC 11 mm Pre-Assembled Clear Polyethylene Snap Cap with PTFE/Silicone (Red/White) Septa, 100/pk
Catalog No. 502104096
Encompass_Preferred
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.
Catalog No. 50-210-4096 Supplier Perkin Elmer US LLC Supplier No. N9303419
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.