Learn More
Cayman Chemical 11-nor-9-carboxy CnabInol 1mg
SDP
Catalog No. 502947447
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid metabolite; a metabolite of cannabinol; intended for research and forensic applications
Catalog No. 50-294-7447 Supplier Cayman Chemical Supplier No. 345921 MG
An analytical reference standard categorized as a phytocannabinoid metabolite; a metabolite of cannabinol; intended for research and forensic applications