Learn More
TARGETMOL CHEMICALS INC 11-O-Galloylbergenin 1MG
SDP
Catalog No. 502879317
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 11-O-Galloylbergenin is isolated from of Mallotus philippinensis with antioxidant and antiplasmodial activities. Purity 99.16%
Catalog No. 50-287-9317 Supplier TARGETMOL CHEMICALS INC Supplier No. T38321MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 11-O-Galloylbergenin is isolated from of Mallotus philippinensis with antioxidant and antiplasmodial activities. Purity 99.16%