Learn More
AdipoGen 11-O-Methylpseurotin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20121M001
Encompass_Preferred
Chemical. CAS: 956904-34-0. Formula: C23H27NO8. MW: 445.5. Isolated from Sporothrix sp. Antibacterial. Selectively inhibits a Hof1 deletion strain.
Catalog No. 50-205-3489