Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11-oxa-2,4,9-triazatridecanoic acid, 8-carboxy-4-[(1,1-dimethylethoxy)carbonyl] (8S) | 97745-69-2 | 99.1% | 474.55 g/mol | C21H38N4O8 | 25 MG
SDP

Catalog No. 5000209050
Encompass_Preferred

Boc-Arg(Boc)2-OH is a Boc-protected derivative of arginine used as an amino-acid building block in peptide synthesis and related organic chemistry applications. It is supplied as a white to off-white solid and is characterized by HPLC, NMR, and MS for identity and purity.

  • High purity: 99.13% (HPLC).
  • White to off-white solid physical form.
  • Molecular formula C21H38N4O8 and molecular weight 474.55 g/mol.
  • Identity confirmed by 1H NMR and mass spectrometry.
  • Suitable as an amino-acid building block for peptide synthesis.

Catalog No. 50-002-09050 Supplier Medchemexpress LLC Supplier No. 25MGHYW088151
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Arg(Boc)2-OH is a Boc-protected derivative of arginine used as an amino-acid building block in peptide synthesis and related organic chemistry applications. It is supplied as a white to off-white solid and is characterized by HPLC, NMR, and MS for identity and purity.

  • High purity: 99.13% (HPLC).
  • White to off-white solid physical form.
  • Molecular formula C21H38N4O8 and molecular weight 474.55 g/mol.
  • Identity confirmed by 1H NMR and mass spectrometry.
  • Suitable as an amino-acid building block for peptide synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.