Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11-Oxomogroside IIa | 5 MG
SDP

Catalog No. 5000419945
Encompass_Preferred

11-Oxomogroside IIa (11-oxomogroside II A1) is a cucurbitane glycoside extracted from the fruits of Siraitia grosvenorii. It has inhibitory effects against the Epstein-Barr virus early antigen (EBV-EA) activation induced by 12-O-tetradecanoylphorbol-13-acetate (TPA), and shows weak inhibitory effects on activation of (+/-)-(E)-methyl-2-[(E)-hydroxyimino]-5-nitro-6-methoxy-3-hexemide (NOR 1), a nitric oxide (NO) donor. It is for research use only and not sold to patients.

  • Extracted from the fruits of Siraitia grosvenorii.
  • Inhibitory effects against Epstein-Barr virus early antigen (EBV-EA) activation.
  • Shows weak inhibitory effects on activation of NOR 1, a nitric oxide (NO) donor.
  • Purity: 99.92%.
  • Molecular weight: 799.00.
  • Chemical formula: C42H70O14.
  • Appearance: Solid.
  • Color: White to light yellow.
  • Classified as terpenoids, triterpenes.

Catalog No. 50-004-19945 Supplier Medchemexpress LLC Supplier No. HYN70415MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

11-Oxomogroside IIa (11-oxomogroside II A1) is a cucurbitane glycoside extracted from the fruits of Siraitia grosvenorii. It has inhibitory effects against the Epstein-Barr virus early antigen (EBV-EA) activation induced by 12-O-tetradecanoylphorbol-13-acetate (TPA), and shows weak inhibitory effects on activation of (+/-)-(E)-methyl-2-[(E)-hydroxyimino]-5-nitro-6-methoxy-3-hexemide (NOR 1), a nitric oxide (NO) donor. It is for research use only and not sold to patients.

  • Extracted from the fruits of Siraitia grosvenorii.
  • Inhibitory effects against Epstein-Barr virus early antigen (EBV-EA) activation.
  • Shows weak inhibitory effects on activation of NOR 1, a nitric oxide (NO) donor.
  • Purity: 99.92%.
  • Molecular weight: 799.00.
  • Chemical formula: C42H70O14.
  • Appearance: Solid.
  • Color: White to light yellow.
  • Classified as terpenoids, triterpenes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.