Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11-Oxomogroside IIIE | 2096516-68-4 | 10 MG
SDP

Catalog No. 5000421141
Encompass_Preferred

11-Oxomogroside IIIE is a cucurbitane triterpene glycoside isolated from Lo Han Kuo (Siraitia grosvenori). It is for research use only.

  • Purity: 99.69%
  • Molecular weight: 961.14
  • Formula: C48H80O19
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, triterpenes
  • Initial source: Plants, cucurbitaceae, Siraitia grosvenorii (Swingle) C. Jeffrey ex Lu et Z. Y. Zhang

Catalog No. 50-004-21141 Supplier Medchemexpress LLC Supplier No. HYN692010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

11-Oxomogroside IIIE is a cucurbitane triterpene glycoside isolated from Lo Han Kuo (Siraitia grosvenori). It is for research use only.

  • Purity: 99.69%
  • Molecular weight: 961.14
  • Formula: C48H80O19
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, triterpenes
  • Initial source: Plants, cucurbitaceae, Siraitia grosvenorii (Swingle) C. Jeffrey ex Lu et Z. Y. Zhang

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.