Learn More
Grainger 11 oz Aerosol Lubricant
Catalog No. 502763773
Encompass_Preferred
The Grainger Approved Lubricant is a multi-purpose, lithium grease ideal for general lubrication of bearings and sliding surfaces. It has an operating temperature range of 0°F to 300°F and contains lithium 12-hydroxystearate thickener, paraffinic base oil, and EP/anti-wear additives. This NLGI Grade 2 grease comes in 14 oz cartridges, 35 lb pails, and 120 lb kegs. It meets military specification MIL-G-18458B-SH. The multi-purpose grease provides corrosion resistance, water resistance, and high load carrying capacity. It can be applied manually or with standard grease guns and automatic lubrication systems.
Catalog No. 50-276-3773 Supplier Grainger Supplier No. 16W546
Add to Cart
Add to Cart
The Grainger Approved Lubricant is a multi-purpose, lithium grease ideal for general lubrication of bearings and sliding surfaces. It has an operating temperature range of 0°F to 300°F and contains lithium 12-hydroxystearate thickener, paraffinic base oil, and EP/anti-wear additives. This NLGI Grade 2 grease comes in 14 oz cartridges, 35 lb pails, and 120 lb kegs. It meets military specification MIL-G-18458B-SH. The multi-purpose grease provides corrosion resistance, water resistance, and high load carrying capacity. It can be applied manually or with standard grease guns and automatic lubrication systems.