Learn More
MSC Super Lube Multi-Purpose Synthetic Lubricant, Aerosol, Food Grade, 11 oz
Catalog No. 501112909
Encompass
Super Lube Multi-Purpose Synthetic Lubricant with Syncolon is a heavy-duty aerosol grease designed for superior wear protection, lubrication, and rust prevention in various applications. It is NSF H1 registered, making it safe for incidental food contact.
  • Prevents rust and corrosion while reducing friction
  • Provides superior anti-wear performance
  • Penetrates and coats surfaces
  • Will not drip, run, or evaporate
  • Extends machinery life, reduces maintenance
  • Remains free-flowing in freezing temperatures
  • Water and saltwater resistant
  • High dielectric strength
  • NSF H1 registered for incidental food contact
  • Kosher certified
Catalog No. 50-111-2909 Supplier MSC Supplier No. 00257048
Super Lube Multi-Purpose Synthetic Lubricant with Syncolon is a heavy-duty aerosol grease designed for superior wear protection, lubrication, and rust prevention in various applications. It is NSF H1 registered, making it safe for incidental food contact.
  • Prevents rust and corrosion while reducing friction
  • Provides superior anti-wear performance
  • Penetrates and coats surfaces
  • Will not drip, run, or evaporate
  • Extends machinery life, reduces maintenance
  • Remains free-flowing in freezing temperatures
  • Water and saltwater resistant
  • High dielectric strength
  • NSF H1 registered for incidental food contact
  • Kosher certified