Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 11-Pentafluorophenoxyundecyltrimethoxysilane

Catalog No. 503126017 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6017 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-312-6017 Supplier Sigma Aldrich Supplier No. SIK440420250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This coupling agent is an hydrophobic ether and gives very good results in the non-covalent immobilization (hydrophobic and pi stacking interactions) of proteins like antibodies.Product of SiKEMIA

Specifications

CAS 944721-47-5
Molecular Formula C20H31F5O4Si
Linear Formula C20H31F5O4Si
Quantity 250 mg
Molecular Weight (g/mol) 458.54
Percent Purity ≥95%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.