Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11(Z),14(Z),17(Z)-eicosatrienoic acid | 17046-59-2 | MFCD00673432 | 99.3% | 306.48 g·mol-1 | C20H34O2 | 5 MG
SDP

Catalog No. 5000263794
Encompass_Preferred

11(Z),14(Z),17(Z)-Eicosatrienoic acid is an all-cis polyunsaturated long-chain fatty acid (C20:3) supplied for research use. It has been reported to support maintenance of functional mitochondria in Saccharomyces cerevisiae (KD115) and is used in biochemical and cellular studies of lipid signaling, membrane composition, and fatty acid metabolism.

  • Unsaturated long-chain fatty acid (C20H34O2), molecular weight 306.48 g·mol-1.
  • High purity (99.3%) suitable for analytical and cellular assays.
  • Liquid appearance, colorless to light yellow, density ≈0.917 g·cm-3.
  • Storage recommendations: pure form -20°C (long-term); in solvent -80°C (short-term).
  • Provided in small research pack sizes for experimental use.

Catalog No. 50-002-63794 Supplier Medchemexpress LLC Supplier No. HY108398B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

11(Z),14(Z),17(Z)-Eicosatrienoic acid is an all-cis polyunsaturated long-chain fatty acid (C20:3) supplied for research use. It has been reported to support maintenance of functional mitochondria in Saccharomyces cerevisiae (KD115) and is used in biochemical and cellular studies of lipid signaling, membrane composition, and fatty acid metabolism.

  • Unsaturated long-chain fatty acid (C20H34O2), molecular weight 306.48 g·mol-1.
  • High purity (99.3%) suitable for analytical and cellular assays.
  • Liquid appearance, colorless to light yellow, density ≈0.917 g·cm-3.
  • Storage recommendations: pure form -20°C (long-term); in solvent -80°C (short-term).
  • Provided in small research pack sizes for experimental use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.