Learn More
Cayman Chemical 11 Z 14 Z 17 ZEIcosatrIen 10mg
SDP
Catalog No. 502966579
Encompass_Preferred
An esterified form of 11(Z),14(Z),17(Z)-eicosatrienoic acid; has been found in the lipid-producing microalgae Nannochloropsis; has been found in diesel-contaminated soils and residential wastewater
Catalog No. 50-296-6579 Supplier Cayman Chemical Supplier No. 1762210 MG
An esterified form of 11(Z),14(Z),17(Z)-eicosatrienoic acid; has been found in the lipid-producing microalgae Nannochloropsis; has been found in diesel-contaminated soils and residential wastewater