Learn More
Cayman Chemical 11(Z)-Eicosenoic Acid, 5561-99-9, 100 mg
SDP
Supplier:  Cayman Chemical 20606100
Encompass_Preferred
One of several monounsaturated 20-carbon fatty acids; combined C20:1 isomers constitute approximately 70% of the total fatty acid pool in jojoba seed oil
Catalog No. NC1462616