Learn More
Cayman Chemical 11(Z)-Eicosenoic Acid, Purity: ≥95%, CAS: 5561-99-9, 1 g
SDP
Catalog No. 502984739
Encompass_Preferred
One of several monounsaturated 20-carbon fatty acids; combined C20:1 isomers constitute approximately 70% of the total fatty acid pool in jojoba seed oil
Catalog No. 50-298-4739 Supplier Cayman Chemical Supplier No. 206061 G
One of several monounsaturated 20-carbon fatty acids; combined C20:1 isomers constitute approximately 70% of the total fatty acid pool in jojoba seed oil