Learn More
Cayman Chemical 11 Z-EIcosenoIc AcId 250mg
SDP
Catalog No. 502982267
Encompass_Preferred
One of several monounsaturated 20-carbon fatty acids; combined C20:1 isomers constitute approximately 70% of the total fatty acid pool in jojoba seed oil
Catalog No. 50-298-2267 Supplier Cayman Chemical Supplier No. 20606250 MG
One of several monounsaturated 20-carbon fatty acids; combined C20:1 isomers constitute approximately 70% of the total fatty acid pool in jojoba seed oil