Learn More
Cayman Chemical 11(Z)-Eicosenoic Acid, Purity: ≥95%, CAS: 5561-99-9, 500 mg
SDP
Catalog No. 502982922
Encompass_Preferred
One of several monounsaturated 20-carbon fatty acids; combined C20:1 isomers constitute approximately 70% of the total fatty acid pool in jojoba seed oil
Catalog No. 50-298-2922 Supplier Cayman Chemical Supplier No. 20606500 MG
One of several monounsaturated 20-carbon fatty acids; combined C20:1 isomers constitute approximately 70% of the total fatty acid pool in jojoba seed oil