Learn More
Cayman Chemical 11(Z)-Vaccenic Acid methyl ester, Purity: ≥98%, CAS: 1937-63-9, 500 mg
SDP
Catalog No. 502984936
Encompass_Preferred
A methyl ester form of cis-vaccenic acid; inhibits sodium influx in mouse brain synaptic plasma membranes, which is correlated with an increase in membrane lipid disordering; has been used as an internal standard for the quantification of cis-vaccenic acid
Catalog No. 50-298-4936 Supplier Cayman Chemical Supplier No. 20693500 MG
A methyl ester form of cis-vaccenic acid; inhibits sodium influx in mouse brain synaptic plasma membranes, which is correlated with an increase in membrane lipid disordering; has been used as an internal standard for the quantification of cis-vaccenic acid