Learn More
Cayman Chemical 11 ZVaccenyl Acetate
SDP
Supplier:  Cayman Chemical 1001010110 MG
Encompass_Preferred
The male-specific mating pheromone of the fruit fly D. melanogaster; acts selectively through the Or67d odorant receptor to control mating behavior in both male and female fruit flies
Catalog No. 50-288-9243