Learn More
Cayman Chemical 11(Z)-Vaccenyl Acetate, Purity: ≥98%, CAS: 6186-98-7, 50 mg
SDP
Catalog No. 502889180
Encompass_Preferred
The male-specific mating pheromone of the fruit fly D. melanogaster; acts selectively through the Or67d odorant receptor to control mating behavior in both male and female fruit flies
Catalog No. 50-288-9180 Supplier Cayman Chemical Supplier No. 1001010150 MG
The male-specific mating pheromone of the fruit fly D. melanogaster; acts selectively through the Or67d odorant receptor to control mating behavior in both male and female fruit flies