Learn More
Burch Bottle & Packaging Inc 110/400 BLACK POLYPROPYLENE CA
Supplier:  Burch Bottle & Packaging Inc 2BP1111MR
Encompass
Stock number 2BP1111MR is a 110/400 injection molded, plastic closure with ribbed sides, matte top, F217 foam and pressure sensitive (PS-22) liners.
Catalog No. NC1778769