Learn More
Savillex Corporation 110 MM CLOSURE - PFA,EACH
SDP
Catalog No. NC1921654
Encompass
•Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C)•External grips on the closure fit 110 mm wrench•Leakproof seal
Catalog No. NC1921654 Supplier Savillex Corporation Supplier No. 60011001
•Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C)•External grips on the closure fit 110 mm wrench•Leakproof seal