Learn More
Abcam 1,10-Phenanthroline, Metalloprotease inhibitor, 25G
Catalog No. 502665979
Encompass_Preferred
MW 180.2 Da, Purity >99%. Membrane permeable chelating agent. Metalloprotease inhibitor. Chelates intracellular heavy metals including zinc, copper and iron. Binds DNA and modulates multiple signaling pathways in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-5979 Supplier Abcam Supplier No. AB14119625G
Add to Cart
Add to Cart
MW 180.2 Da, Purity >99%. Membrane permeable chelating agent. Metalloprotease inhibitor. Chelates intracellular heavy metals including zinc, copper and iron. Binds DNA and modulates multiple signaling pathways in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement