Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HistoPrep™ Acetone, Fisherbrand™
SDP

Catalog No. 11002200 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-200 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-200
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

A cleaning solvent and fixative.

  • Can be used as a fixative to preserve tissue integrity by breaking hydrogen bonds, removing and replacing water, and precipitating proteins.
  • For common uses including removing mounting media and mordant.

Chemical Identifiers

CAS 67-64-1

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 1 gal.
Percent Purity 99%
Grade ACS

Made in the USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.