Learn More
Sino Biological Recombinant Human CD20 Protein (aa 213-297, His Tag), HPLC-verified
Supplier:  Sino Biological 11007H07H220
Encompass_Preferred
A DNA sequence encoding the human CD20 (NP_068769.2) (Glu213-Pro297) was expressed, with an N-terminal polyhistidine tag.
Catalog No. 50-161-5995