Learn More
Sino Biological Recombinant Human SerpinI2 Protein (His Tag)
Supplier:  Sino Biological 11008H08H100
Encompass_Preferred
A DNA sequence encoding the human SERPINI2 (NP_006208.1) (Met 1-Leu 405) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5998