Learn More
Sino Biological Recombinant Human Cyclin A1 Protein (His Tag)
Supplier:  Sino Biological 11013H07B100
Encompass_Preferred
A DNA sequence encoding the full length of human CCNA1 isoform a (NP_003905.1) (Met 1-Gln 465) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6006