Learn More
Sino Biological Recombinant Human HK3 Protein (His & GST Tag)
Supplier:  Sino Biological 11016H20B50
Encompass_Preferred
A DNA sequence encoding the human HK3 (P52790) (Met 1-Val 923) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-6012