Learn More
Sino Biological Recombinant Human GPR56 Protein (His Tag)
Supplier:  Sino Biological 11017H08H100
Encompass_Preferred
A DNA sequence encoding the human GPR56 isoform b (NP_958933.1) extracellular domain (Met 1-Val 342) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6013