Learn More
Sino Biological Recombinant Human IL-22BP Protein (His & hFc Tag)
Supplier:  Sino Biological 11025H03H50
Encompass_Preferred
A DNA sequence encoding the human IL22RA2 isoform 2 (NP_851826.1) (Met 1-Pro 231) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6026