Learn More
Sino Biological Recombinant Human CD5 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11027H08H100
Encompass_Preferred
A DNA sequence encoding the human CD5 (NP_055022.2) extracellular domain (Met 1-Pro 372) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6029