Learn More
Sino Biological Recombinant Human CD7 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11028H08H100
Encompass_Preferred
A DNA sequence encoding the human CD7 (NP_006128.1) extracellular domain (Met 1-Pro 180) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6031