Learn More
Sino Biological Recombinant Human CD24 Protein (hFc Tag), HPLC-verified
Catalog No. 501616036
Encompass_Preferred
A DNA sequence encoding the human CD24 (NP_037362.1) (Met 1-Gly 59) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6036 Supplier Sino Biological Supplier No. 11030H02H100
Add to Cart
Add to Cart
A DNA sequence encoding the human CD24 (NP_037362.1) (Met 1-Gly 59) was fused with the Fc region of human IgG1 at the C-terminus.