Learn More
Sino Biological Recombinant Human CD8 beta Protein, HPLC-verified
Supplier:  Sino Biological 11031HCCH100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD8B (P10966-1) (Met1-Pro170) was expressed with six amino acids (LEVLFQ) at the C- terminus.
Catalog No. 50-161-6040