Learn More
Sino Biological Recombinant Human PHKG1 Protein (GST Tag)
Supplier:  Sino Biological 11034H09B50
Encompass_Preferred
A DNA sequence encoding the human PHKG1 (NP_006204.1) (Met 1-Tyr 387) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-6046