Learn More
Sino Biological Human INS Protein
Supplier:  Sino Biological 11038HNAY100
Encompass_Preferred
A DNA sequence encoding human insulin B chain (P01308)(Phe25-Lys53) and human insulin A chain (P01308)(Gly90-Asn110) was expressed with a linker AAK.
Catalog No. 50-161-6047