Learn More
Sino Biological Recombinant Human N-Cadherin Protein (His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 11039H03H100
Encompass_Preferred
A DNA sequence encoding the human CDH2 (NP_001783.2) (Met 1-Ala 724) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6049