Learn More
Sino Biological Recombinant Human N-Cadherin Protein (His Tag)
Supplier:  Sino Biological 11039H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CDH2 (NP_001783.2) precursor (Met 1-Ala 724) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6053