Learn More
Sino Biological Recombinant Human SP-D/SFTPD Protein (His Tag)
Supplier:  Sino Biological 11041H08H100
Encompass_Preferred
A DNA sequence encoding the human SFTPD (NP_003010.4) precursor (Met 1-Phe 375) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6057