Learn More
Sino Biological Recombinant Human CD16b/FCGR3B/Fc gamma RIIIB Protein (NA2 allotype, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11046H27HB20
Encompass_Preferred
A DNA sequence encoding the human FCGR3B (O75015.2) (Met1-Ser200) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus (AVI-his). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-161-6072