Learn More
Sino Biological Recombinant Rabbit IgG Protein (185 Thr/Ala, Asn 284/Ser)
Supplier:  Sino Biological 11047TNAH100
Encompass_Preferred
A DNA sequence encoding the rabbit IgG Fc region (P01870) (Ser 101-Lys 323, 185Thr/Ala, Asn 284/Ser) was expressed and purified.
Catalog No. 50-161-6078