Learn More
Sino Biological Influenza A H5N1 (A/Anhui/1/2005) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11048V08B100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus (A/Anhui/1/2005(H5N1) (ABD28180.1) hemagglutinin (Met 1-Gln 527,Ala 150Val) with cleavage site mutated (RERRRKR→TETR, HA1+HA2, uncleaved) was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6083