Learn More
Sino Biological Influenza A H5N1 (A/Anhui/1/2005) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11048V08H2100
Encompass_Preferred
A DNA sequence encoding the the N-terminal segment (Met 1-Pro 337) of Influenza A virus H5N1 hemagglutinin (A/Anhui/1/2005 (H5N1) (ABD28180.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6089