Learn More
Sino Biological Influenza A H5N1 (A/Anhui/1/2005) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11048V08H4100
Encompass_Preferred
A DNA sequence encoding the native extracellular domain of Influenza A virus (A/Anhui/1/2005 (H5N1) (ABD28180.1) hemagglutinin (Met 1-Gln 530) (Native, HA1+HA2, cleaved) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6092